Biox Inoculating Loops and Needles
Loop, 1 µL, Blue
Catalog # 75799-952
Supplier: TRADEWINDS DIRECT INC
Supplier #: BX65-0001
Jump to:
Product Details & Documents
Innoculating loops and needles are smooth and flexible for uniform and smooth streaking, without damaging the gel surface.
- Sterile
- Polystyrene
- Other end of the loops can be used as a needle
- Free from lubricants, oils, and electrostatic charges
Needles are suitable for removal of single colony specimens and the opposite end of each loop can be used as a needle.
Specifications
- Color:Blue
- Description:Loop, 1 µL