Biox Transfer Pipettes
160 mm, Sterile, To 3 mL, Individually Wrapped
75799-908
: BX30-0138A1-1
Transfer Pipettes are perfect for quick and safe liquid transfer.
- Disposable
- Polyethylene construction
- Uniform drop size
- 100% inert for accurate sampling
Polyethylene pipettes are a great alternative to pasteur pipettes or rubber bulbs.
- Sterility:Sterile
- Length:160 mm
- Packed:Individually Wrapped
- Graduations:To 3 mL