Orders LinkContactUsLinkComponent
Biox Cardboard Freezer Boxes
100, 5 cm (2")
  75780-314
 :  BX90-2200
Biox Cardboard Freezer Boxes
  75780-314
Biox Cardboard Freezer Boxes
Biox Cardboard Freezer Boxes
100, 5 cm (2")
$9,111.85
Case (100UN)
Premium cardboard freezer boxes with a water-resistant coating.
  • Chemically resistant to alcohols and mild organic solvents
  • Temperature range: –196 to 121°C (–320 to 250 °F)
  • Accommodates 0.5 ml, 1.5 ml, or 2.0 ml microtubes
  • Suitable for liquid nitrogen storage
Boxes feature a water-resistant coating for improved durability.
  • No. of Holes:
    100
  • Height:
    5 cm (2")