BBL™ Penicillinase, BD Biosciences
Catalog # 90000-426
Supplier: BECTON DICKINSON DE MEXICO, S. MX
CAS Number:
Specifications
- Descripción:BBL™ Penicillinase Concentrate
- Tamaño:100 mL
- Referencia:90000-426
- Nº de proveedor:211899
Specifications
About this item
This enzyme is specifically used for the inactivation of penicillin, for increasing the susceptibility of Gram-negative bacteria to antibiotics.
- Interferes with the antigen-antibody reaction
- Increases susceptibility of Gram-negative bacteria to antibiotics
Penicillinases provide multi-resistance to antibiotics such as penicillins, cephamycins, and carbapenems. Through hydrolysis, penicillinases break the molecular structure that all these antibiotics have in common.