Orders LinkContactUsLinkComponent
Inoculum Broth, BD
Inoculum Broth, BD
Certificates
Inoculum Broth, BD
2 Options Available Below
Product Details & Documents
Inoculum Broth is used for preparing the inoculum of lactobacilli and other microorganisms used in microbiological assays of vitamins and amino acids.
  • Used for cultivating lactobacilli
  • For preparing the inoculum for microbiological assays
  • Peptone provides nitrogen and amino acids
  • Beige, homogenous appearance when dehydrated
Inoculum Broth is the recommended medium for preparation of lactobacilli inoculum for vitamin assays.
Product Family Options
Información del producto
  • Size
    Availability
    Price
  • 90003-490
    B12 Inoculum Broth
    B12 Inoculum Broth
    100 g
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (100g)
    Each (100g) - $4,580.00
    $4,580.00
    Each (100g)
       $4,580.00
    Supplier Number: 254210
    El artículo requiere control de temperatura para almacenamiento y entrega con cargos adicionales. No es elegible para devolución debido a preocupaciones de seguridad y calidad. Considere los requisitos antes de comprar.
  • 90000-366
    Difco™ Micro Inoculum Broth
    Difco™ Micro Inoculum Broth
    500 g
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (500g)
    Each (500g) - $10,880.00
    $10,880.00
    Each (500g)
       $10,880.00
    Supplier Number: 211813