Zipper Bags
Clearzip® Lock top bags with white block, 4×6"
Catalog # 470150-359
Supplier: INTERNATIONAL PLASTICS CP
Número de proveedor #: CZW20406
Jump to:
Product Details & Documents
Clearzip® Lock Top Bags are regular duty clear plastic zipper bags that feature a 2 Mil white block area for organizing, packaging, and storage items.
- Made from low density polyethylene
- Reclosable and resealable
- White block accepts rubber stamp, marker, grease pen, or ballpoint pen
Ideal for industrial, food service and healthcare applications.
Certifications: Compilance with FDA/USDA requirements for use in food packaging and storage applications.
Especificaciones
- Description:Clearzip® Lock top bags with white block, 4×6"