Microbank Cryoblock, Pro-Lab Diagnostics
Microbank™ Cryoblock
Catalog # 75784-932
Supplier: PRO-LAB DIAGNOSTIC
Número de proveedor #: PL.155-1
Jump to:
Product Details & Documents
A 20 well, insulated cryoblock is available which enables the operator to remove a number of Microbank™ vials from the freezer for sub culture without risk of the culture beads defrosting.
The cryoblock is insulated with a polystyrene shield.
Especificaciones
- Description:Microbank™ Cryoblock