Orders LinkContactUsLinkComponent
VWR® Pean Hemostatic Forceps
Supplier:   VWR International
VWR® Pean Hemostatic Forceps
Supplier:  VWR International
Certificates
VWR® Pean Hemostatic Forceps
VWR® Pean Hemostatic Forceps
3 Options Available Below
Product Details & Documents
VWR® Pean Hemostatic Forceps are constructed of surgical grade stainless steel
  • Passivated
  • Box-lock joint
  • Surgical grade
  • Rockwell Hardness (HRC): 40–45
  • Fully autoclavable
  • Compatible with Gamma/ETO/Autoclavable sterilization methods
These instruments are compatible with a variety of sterilization methods, and are available in an assortment of lengths and jaw types.
Product Family Options
Información del producto
  • Material
    Length
    Jaw Type
    Curvature
    Availability
    Price
  • 10806-182
    VWR® Pean Hemostatic Forceps
    VWR® Pean Hemostatic Forceps
    Stainless steel, surgical grade
    159 mm (6.25")
    Smooth
    Straight
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $886.08
    $886.08
    Log in Para disponibilidad
    Paquete de 12 (1UN)
    Paquete de 12 (1UN) - $965.83
    $965.83
    Each (1UN)
       $886.08
    Paquete de 12 (1UN)
       $965.83
  • 10806-188
    VWR® Pean Hemostatic Forceps
    VWR® Pean Hemostatic Forceps
    Stainless steel, surgical grade
    184 mm (7.25")
    Full serrated
    Curved
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $888.82
    $888.82
    Log in Para disponibilidad
    Paquete de 12 (1UN)
    Paquete de 12 (1UN) - $968.82
    $968.82
    Each (1UN)
       $888.82
    Paquete de 12 (1UN)
       $968.82
  • 10806-200
    VWR® Pean Hemostatic Forceps
    VWR® Pean Hemostatic Forceps
    Stainless steel, surgical grade
    305 mm (12")
    Full serrated
    Curved
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $1,189.73
    $1,189.73
    Log in Para disponibilidad
    Paquete de 12 (1UN)
    Paquete de 12 (1UN) - $1,296.81
    $1,296.81
    Each (1UN)
       $1,189.73
    Paquete de 12 (1UN)
       $1,296.81