VWR® Vial Decapper Pliers
VWR® Vial Decapper Pliers, 8 mm
Catalog # 10806-366
Supplier: VWR International
Jump to:
Product Details & Documents
VWR® Vial Decapper Pliers are designed to easily remove aluminum crimp tops from vials and bottles.
- Passivated
- Surgical-grade stainless steel construction
- Compatible with Gamma/ETO/Autoclavable sterilization methods
Available in single- or double-action formats, all pliers are smooth and simple to operate. Durable, high-quality construction ensures consistent, dependable performance.
Overall length: 159 mm (6.25")
Documents
Certificates
Buscar por número de loteEspecificaciones
- Description:VWR® Vial Decapper Pliers
- Seal Size:8 mm