Orders LinkContactUsLinkComponent
VWR® Dissection Needle with Plastic Holder
VWR® Dissection Needle with Plastic Holder
VWR® Dissection Needle with Plastic Holder, 14.4 cm (5.68"), Sharp, Straight
Catalog # 10806-326
Supplier:   VWR International
VWR® Dissection Needle with Plastic Holder
Catalog # 10806-326
Supplier:  VWR International
VWR® Dissection Needle with Plastic Holder
VWR® Dissection Needle with Plastic Holder
VWR® Dissection Needle with Plastic Holder, 14.4 cm (5.68"), Sharp, Straight
Price
$20.92
Log in Para disponibilidad
Each (1UN) Paquete de 12 (1UN)
Product Details & Documents
Sharp-pointed, straight needles feature a plastic holder.
  • Passivated
  • Compatible with Gamma/ETO/Autoclavable sterilization methods
Available in straight or bent formats, all devices have a needle length of 45mm (1.75").
Especificaciones
  • Description:
    VWR® Dissection Needle with Plastic Holder
  • Length:
    14.4 cm (5.68")
  • Curvature:
    Straight
  • Tip Style:
    Sharp