Slide Tray Cabinet
Slide Trays
Catalog # 470157-670
Supplier: BEL-ART PRODUCTS SE
Número de proveedor #: H44581-0000_SE
Jump to:
Product Details & Documents
Store your slides securely with a minimal surface footprint.
- Store slides horizontally for the best care of whole mounts
- Clear door allows for internal viewing
Our polypropylene microscope slide tray cabinet holds up to 240 slides total. It can accommodate 12 horizontal slide trays in all. The clear acrylic door slides vertically. Slide trays are available separately.
Especificaciones
- Description:Slide Trays