Boreal Science Square Cover Glasses
Catálogo # 470145-878
Proveedor: C & A SCIENTIFIC CO INC
Especificaciones
- Numbering:#2
- Descripción:Cover glasses, square
- l×a:18×18 mm
- Grosor:0.18 - 0.25 mm
- Referencia:470145-878
- Nº de proveedor:94-18182
Especificaciones
Acerca de este artículo
Traditional Style Coverslips.
- Quality slide making supplies
- Traditional style coverslips
Precisely cut cover glasses for your slide making needs. Available in different thicknesses and dimensions. Quantity 75–100.