Disposable Samplers, MultiDispo, Burkle
10821-261
- Ø:2.5 cm (0.98")
- Volumen:100 ml
- Descripción:MultiDispo, Standard
- Longitud:50 cm (19.6")
- Environmentally Preferable:Low Manufacturing Impact
- Certificación ambiental:ISO 14001 Certificate
- Referencia:10821-261
- Sterile:Non-Sterile
- Nº de proveedor:5393-4440
Disposable zone samplers for taking samples from multiple layers of the sample material.
- HDPE
- Clean room manufactured
- Optionally sterilised by gamma rays
- Easy emptying through the opening in the handle (Ø 14 mm)
Suitable for free-flowing powders and small-grain granulates. The representative all-layer sample can be emptied directly through the open handle into a sample container.