Disposable Samplers, DispoPicker, Burkle
Catálogo # 10821-331
Proveedor: BURKLE INC
Especificaciones
- Ø:2.1 cm (0.8")
- Volumen:100 ml
- Descripción:DispoPicker, Sterilized
- Longitud:50 cm (19.6")
- Referencia:10821-331
- Sterile:Sterile
- Nº de proveedor:5393-5551
Especificaciones
Acerca de este artículo
DispoPicker is ideally suited for cross-section samples.
- HDPE
- Clean room manufactured
- Optionally sterilised by gamma rays
- Cross contamination is completely eliminated
Disposable sampler for powders and small-grain bulk materials. Because the tip is closed, the sample material cannot trickle back out of the sample lance.