Disposable Samplers, ViscoDispo, Burkle
Catalog # 10821-339
Supplier: BURKLE INC
Specifications
- Ø:2.1 cm (0.8")
- Volumen:200 ml
- Descripción:ViscoDispo, Sterilized, Pack of 20
- Longitud:100 cm (39.3")
- Referencia:10821-339
- Sterile:Sterile
- Nº de proveedor:5393-3331
Specifications
About this item
The ViscoDispo disposable sampler can be used to take samples of high-viscosity media up to 100.000 mPas.
- HDPE
- Without seals
- Clean room manufactured
- Optionally sterilised by gamma rays
- Cross contamination is completely eliminated
The ergonomically shaped handle and the very stable drawbar make it easy to draw even viscous substances.
Ordering information: Each Unit contains 20 packs.