Demonstration Transformer Kit
Transformer accessory kit 1
Catalog # 470219-516
Supplier: EISCO SCIENTIFIC LLC
Número de proveedor #: PH0849AK1
Jump to:
Product Details & Documents
The Demonstration Transformer Kit is a safe and convenient system for AC and DC experiments up to 0.5 amps.
- For AC and DC Experiments up to 0.5A
- Large-Scale Transformer
- Ideal for the Classroom
- Intermediary Outputs
- Safe and Convenient
The large-scale transformer is ideal for the classroom.
The coils are fitted with intermediary outputs that provide a variety of transformation ratios.
: The accessory kit demonstrates how step down transformers can produce currents that are large enough to weld metals or boil water.
Especificaciones
- Description:Transformer accessory kit 1