VWR® Aluminum Weighing Dish
10806-406
- Capacidad:0.025 L (0.85 oz.)
- Descripción:VWR® Aluminum Weighing Dish
- Dimensiones:15.9 H x 44.5 O.D. mm (0.625 x 1.75")
- Referencia:10806-406
The VWR® Aluminum Weighing Dish is made from a one-piece ASTM standard (B209-10) aluminum sheet, spun to shape.
- Passivated
- Compatible with Gamma/ETO/Autoclavable sterilization methods
Inverted covers allow for easy stacking and storage.