Orders LinkContactUsLinkComponent
Viable Bean Seeds
Viable Bean Seeds
Viable Bean Seeds
Viable Bean Seeds
Typical Economic Crops.
Whether you are showing germination, root growth or testing environmental factors beans are an easy to grow plant exhibiting typical dicot structures. Dried beans are also ideal to use as counters, or for experiments of manipulation and dexterity. Extra seeds can remain viable for up to 3 years with careful storage.
 : Where a Count of Seeds is indicted, the Quantity is an estimate based on Weight and may vary slightly.
Product Family Options
  • Description
    Size
  • Kentucky wonder bean seeds
    2 oz.
     : 805
  • Lima bean seeds
    8 oz.
    Paquete de 1
    Paquete de 1 - $109.26
    $109.26
    Paquete de 1
       $109.26
     : 807*8 OZ.