Lab Burner
Lab burner (supplied without gas cartridge)
Catalog # 470019-572
Supplier: EISCO SCIENTIFIC LLC
Supplier #: CH2019001
Jump to:
Product Details & Documents
These laboratory burners are compact with a powerful and concentrated flame.
- Made of brass and stainless steel
Soot-free use and adjustable flame control. The burners screw into a cartridge to guarantee a leak free installation.
Burner has separate adjustments for regulating gas and air intake. Flame top: 1¼" diameter, burner unit: 2¼"×2⅔".
Documents
VWR GHS Compliance Letter (US)
Additional Product Details
Specifications
- Description:Lab burner (supplied without gas cartridge)