Accessories for KIMBLE® ULTRA-WARE® 47 mm Microfiltration Assembly with GL45 Style Bottle, DWK Life Sciences
Catalog # 10135-800
Supplier: DWK Life Sciences (KIMBLE)
Specifications
- Descripción:PBT GL32 cap, 20 mm opening
- Referencia:10135-800
- Nº de proveedor:410170-3220