Compact Sling Psychrometers
Compact Sling Psychrometer, 21 cm (8.3"), –5...+50 °C, 1 °C, 1 °C
Catalog # 470200-002
Supplier: BEL-ART PRODUCTS SE
Número de proveedor #: B61503-0000
Jump to:
Product Details & Documents
Measure relative humidity with weather bureau-type wet and dry bulb thermometers. Pocket-sized psychrometer is perfect for student use, as it presents no mercury hazards.
- Two organic liquid filled thermometers with white back glass are housed in a durable case
- Available in both Farenheit and Celsius
- Replacement wicks available
Especificaciones
- Temperature Range:–5...+50 °C
- Description:Compact Sling Psychrometer
- Length:21 cm (8.3")
- Graduation Interval:1 °C
- Temperature Accuracy:1 °C