Pocket Hardness Points Set
Pocket Hardness Points Set, 7
Catalog # 470014-828
Supplier: Avantor
Jump to:
Product Details & Documents
Students determine a mineral's hardness between five and 10 on Mohs' scale.
- Students Determine a Mineral's Hardness
- Seven Cut Minerals Mounted on Four Rods
- Rods Stamp with Each Mineral's Hardness
The neat little case tucks into a pocket, so students have an accurate gauge of harder minerals ready. Seven cut minerals, representing Mohs’ hardness measures of 5, 6, 7, 8, 8 1/2, 9, and 10, are permanently mounted on the ends of four pen-size aluminum rods.
The rods are stamped with each mineral’s hardness.
Especificaciones
- Description:Pocket Hardness Points Set
- No. of Samples:7