Ward's® Bookstore Dissection Kit
Bookstore Dissection Kit
Catalog # 470220-350
Supplier: DR INSTRUMENTS INC. SE
Número de proveedor #: DRI78651
Jump to:
Product Details & Documents
Specially built starter set of dissection tools.
- High-quality stainless steel instruments
- Ideal for college biology freshmen
- Supplied with a zippered case for convenient storage
The Bookstore Dissection Set is designed to support all types of college level dissection. The mix of tools allows users to have exactly the necessary item at hand for any basic dissection. The instruments are high-quality stainless steel and will provide years of service. They conveniently store under loops in the zippered case.
Packaging: Please see datasheet for contents.
Caution: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.
Documents
Bookstore Dissection Kit
Hoja de datos
Especificaciones
- Description:Bookstore Dissection Kit