Thermometer, Red
Thermometer, Red, −30...+110 °C (−20...+230 °F)
Catalog # 470193-484
Supplier: PRECISION MOULDED POLYMERS LTD
Número de proveedor #: AM1060.HB006.00000
Jump to:
Product Details & Documents
This set of 6 Science Kit thermometers is ideal for your classroom. They are easy to read and individually wrapped.
Especificaciones
- Description:Thermometer, Red
- Range:−30...+110 °C (−20...+230 °F)