Orders LinkContactUsLinkComponent
3" Test Sieves, W.S. Tyler™
3" Test Sieves, W.S. Tyler™
Supplier:   W.S. Tyler
3" Test Sieves, W.S. Tyler™
Supplier:  W.S. Tyler
Certificates
3" Test Sieves, W.S. Tyler™
3" Test Sieves, W.S. Tyler™
6 Options Available Below
Product Details & Documents
These test sieves are available with stainless or brass frame and stainless cloth.
  • Depth to cloth: 1-1/4"
Product Family Options
Información del producto
  • Ø
    Height
    Mesh Material
    Mesh Size
    USA Series
    Frame Material
    Availability
    Price
  • 3"
    44.5 mm (1¾")
    Stainless
    75 µm
    #200
    Stainless steel
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $3,498.04
    $3,498.04
    Each (1UN)
       $3,498.04
    Supplier Number: 1310
  • 3"
    44.5 mm (1¾")
    Stainless
    150 µm
    #100
    Brass
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $2,504.77
    $2,504.77
    Each (1UN)
       $2,504.77
    Supplier Number: 1555
  • 3"
    44.5 mm (1¾")
    Stainless
    250 µm
    #60
    Stainless steel
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $2,983.50
    $2,983.50
    Each (1UN)
       $2,983.50
    Supplier Number: 1303
  • 3"
    44.5 mm (1¾")
    Stainless
    355 µm
    #45
    Stainless steel
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $2,983.50
    $2,983.50
    Each (1UN)
       $2,983.50
    Supplier Number: 1301
  • 3"
    44.5 mm (1¾")
    Stainless
    850 µm
    #20
    Stainless steel
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $2,983.50
    $2,983.50
    Each (1UN)
       $2,983.50
    Supplier Number: 1296
  • 3"
    44.5 mm (1¾")
    Stainless
    1 mm
    #18
    Stainless steel
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $2,983.50
    $2,983.50
    Each (1UN)
       $2,983.50
    Supplier Number: 1295