ContactUsLinkComponent
Corning® XT Starter Ice-free Cooler, Corning
Corning® XT Starter Ice-free Cooler, Corning
Catálogo # 75779-828
Proveedor:  CORNING CA
undefined
Corning® XT Starter Ice-free Cooler, Corning
Catálogo # 75779-828
Proveedor:  CORNING CA
Número de proveedor:  432014
Especificaciones
  • Descripción:
    XT Starter, complete, purple XT holder and XT cooling core
  • Dimensiones:
    16.2×12.1×6.1 cm
  • Referencia:
    75779-828
  • Nº de proveedor:
    432014
Especificaciones
Acerca de este artículo
An open-platform ice-free cooler that accommodates most CoolRack® and CoolSink® modules.
  • Consists of a base, cooling core and a lid
  • Includes a 1 to 8 °C temperature indicator for visual assurance of proper temperature
  • To use with most Corning CoolRack® and CoolSink® Modules
  • The internal cooling or freezing cores provide 0.5 to 4 °C for up to 4 hours
  • One module capacity
  • Purple color
Low profile and small footprint make it ideal for use in the hood, keeping samples cold (0.5 to 4.0°C) up to four hours. 1 to 8 °C temperature indicator provides visual assurance of temperature performance. To extend the cooling duration, keep an additional XT Cooling Core in the freezer and rotate the Cores as needed.