Orders LinkContactUsLinkComponent
Micro Titertubes, Thermo Scientific
Micro Titertubes, Thermo Scientific
1.2 ml, PP, Natural, Micro Titertube, Robotic Rack
Catalog # 46300-506
Número de proveedor #:  RC-845-TP
Micro Titertubes, Thermo Scientific
Catalog # 46300-506
Micro Titertubes, Thermo Scientific
1.2 ml, PP, Natural, Micro Titertube, Robotic Rack
Price
$1,610.00
Log in Para disponibilidad
Paquete de 960 Case de 5 (960UN)
Product Details & Documents
Micro Titertubes offer additional volume capacity for sample dilution and/or mixing prior to transfer into micro titer plates.
  • Pre-racked in an 8×12 format
  • Available plain or with frosted writing areas
  • Autoclaved
  • Sealed and frozen for storage
Ideal for a wide variety of laboratory procedures such as HTLV-III testing, RIA, EIA, PCR, pharmaceutical quality control, blood bank sample freezer storage or transport of specimens and reagents.
Especificaciones
  • Capacity:
    1.2 ml
  • Color:
    Natural
  • Description:
    Micro Titertube
  • Material:
    PP
  • Packaging:
    Robotic Rack