Replacement Gas Filters for Restek Super Clean® Gas Filter Kits, Restek
Catalog # RK22020
Supplier: RESTEK CORP
Specifications
- Descripción:Replacement Triple Gas Filter (removes oxygen, moisture, and hydrocarbons)
- Referencia:RK22020
- Nº de proveedor:22020
Specifications
About this item
Ensure high-purity output in these gas filters with 99.9999% pure gas (at a max flow of 2 L/min) with the use of these filter kits.
- Designed for easy, hassle-free cartridge changes
- Glass inside to prevent diffusion; polycarbonate housing outside for safety
All traps measure 10⁵/₈×1³/₄" (27×4.4 cm).