Interlock Vials, Restek
Interlock Vial, Thread, 0.30 ml, 12×32 mm, Glass, Clear, No, 9 mm
Catalog # 10058-772
Supplier: RESTEK CORP
Número de proveedor #: 22433
Jump to:
Product Details & Documents
Interlock limited volume vials offer the performance and convenience of a one-piece microvial for a fraction of the cost.
- Clear or amber color
- Screw-thread, crimp-top, or snap-ring formats
- Glass fabrication
A 300 µl insert is fused into a 12×32 mm clear or amber glass vial. This one-piece design reduces the potential for contamination.
Especificaciones
- Cap included:No
- Cap Size:9 mm
- Capacity:0.30 ml
- Color:Clear
- Description:Interlock Vial, Thread
- Material:Glass
- Diameter ×H:12×32 mm