ContactUsLinkComponent
Cyclo Inlet Liner for Agilent GCs Equipped with Split/Splitless Inlets, Restek
Cyclo Inlet Liner for Agilent GCs Equipped with Split/Splitless Inlets, Restek
  RK20707-214.5
Cyclo Inlet Liner for Agilent GCs Equipped with Split/Splitless Inlets, Restek
  RK20707-214.5
 :  20707-214
  • Ext.Ø:
    6.3 mm
  • Volumen:
    0.634 ml
  • Descripción:
    Cyclo Inlet Liner
  • Matriz:
    Borosilicate glass
  • Referencia:
    RK20707-214.5
  • Para ser utilizado con:
    AGLS
  • I.D.×L:
    4.0×78.5 mm
  • Treatment:
    Siltek deactivated
  • Nº de proveedor:
    20707-214.5
Use of these liners helps ensure an inert GC flow path for higher sensitivity, accuracy, and reproducibility.
  • Consistent quality with exceptional batch-to-batch uniformity
  • Low to zero bleed or background contamination
Superior coverage is delivered even with highly active compounds.