Sample Vial Racks, 2.0 ml, Restek
Polypropylene Storage Rack, 50 vials, 5 x 10, 12 x 32 mm
10058-822
: 22856
Sample vial racks feature alphanumeric indexing for easier vial identification.
- Racks can be stacked for efficient storage
- Fits most 2.0 ml, 12×32 mm vials
- Polypropylene fabrication
- Description:Polypropylene Storage Rack
- Size:50 vials, 5 x 10
- For Tubes:12 x 32 mm