ContactUsLinkComponent
Certified Autosampler Vial Kits, Certified 2.0 ml Screw-Thread Vials, Restek
  10058-792
Certified Autosampler Vial Kits, Certified 2.0 ml Screw-Thread Vials, Restek
  10058-792
 :  22789
  • Tapa incluida:
    Si
  • Septo:
    PTFE/silicone, bonded, pre-slit
  • Capacidad:
    2.00 ml
  • Color:
    Clear
  • Descripción:
    Certified Autosampler Vial Kit, Assembled Vial with Marking Spot and Cap
  • Referencia:
    10058-792
  • Nº de proveedor:
    22789
Certified 2.0 ml screw-thread vial kits are available for optimal use with autosamplers.
  • Certificates enclosed for each kit
  • Clear or amber borosilicate glass fabrication
  • Closures with pre-inserted PTFE/silicone septa
Each kit is sealed for ensured integrity. Certificate parameters include vial dimensions and chemical test results.