Precleaned Volatile Organic Analyte, VOA Sampling Vials, Restek
Supplier: RESTEK CORP
Jump to:
About this item
Precleaned volatile organic analyte sampling vials are cleaned and assembled before delivery, and are lot traceable for guaranteed quality.
- Screw-thread or vial format
- Clear or amber color
- Glass fabrication
- 24 mm orifice
Volume capacity ranges from 20 to 60 ml depending on model. These vials are cleaned to U.S. EPA Protocol B specifications.
3 Options Available Below
Product Details & Documents
Precleaned volatile organic analyte sampling vials are cleaned and assembled before delivery, and are lot traceable for guaranteed quality.
- Screw-thread or vial format
- Clear or amber color
- Glass fabrication
- 24 mm orifice
Volume capacity ranges from 20 to 60 ml depending on model. These vials are cleaned to U.S. EPA Protocol B specifications.
Product Family Options
Product Information
- CapacityColorCap includedCap SizeSeptumThicknessAvailabilityPrice
- 10803-256Precleaned VOA VialsPrecleaned VOA Vials40.00 mlClearYes24-400PTFE/silicone0.125"Log in for availabilityPaquete de 72$4,493.46Supplier Number: 21796
- 10854-098Precleaned VOA VialsPrecleaned VOA Vials40.00 mlAmberYes24-400PTFE/silicone0.125"Log in for availabilityPaquete de 72$5,563.39Supplier Number: 21797
- 10854-102Precleaned VOA VialsPrecleaned VOA Vials20.00 mlClearYes24-400PTFE/silicone0.125"Log in for availabilityPaquete de 72$5,392.16Supplier Number: 21798