Replacement Inlet Seals for Agilent GCs, Restek
RK21318
- Ø int.:0.8 mm
- Descripción:Replacement Inlet Seal
- Referencia:RK21318
- Nº de proveedor:21318
The inlet seal at the base of the Agilent 5890/6890/7890 GC injection port contacts the sample and must be changed frequently to prevent adsorption of active compounds.
- All seals include washers
In addition, septum fragments and sample residue accumulate on the disk surface, requiring disk replacement
Note: The 1.2 mm inlet seal is recommended when installing two columns using a two-hole Vespel®/graphite ferrule.
To reduce breakdown and adsorption of active compounds, use Siltek® or gold-plated seals.
: Note: The 1.2 mm inlet seal is recommended when installing two columns using a two-hole Vespel®/graphite ferrule.