Aluminum Seals with Septa, 20 mm, preassembled, Restek
10058-686
- Tipo:Crimp-Top
- Color:Silver
- Descripción:Seals w/Septa
- Matriz:Aluminum
- Referencia:10058-686
- Nº de proveedor:21761
- Orifice:20 mm
Restek's aluminum seals are durable for reliable pressured seals to a predefined release level.
- Pressure seal
- Sturdy aluminum construction
- Economical
- Crimp-top design
Aluminum pressure release seals cover 20-millimeter orifices to create effective seals.
: Pressure release point: 3.0 bar ± 0.5 bar.