Ceramic Scoring Wafer, Restek
Ceramic Scoring Wafers
RK-20116
: 20116
Four straight scoring edges for cutting fused silica tubing and four serrated edges for cutting MXT metal capillary columns.
- Marked increments for easily measured scoring
Crafted from durable ceramic material.
- Description:Ceramic Scoring Wafers