Ward's® Mineral Thin Sections Set
Mineral Thin Sections Set, 40
Catalog # 470024-866
Supplier: Avantor
Jump to:
Product Details & Documents
Forty mineral thin sections for Petrography Studies
- Pure Mineral samples
- Hand Made by Wards® staff Geologists
Our excellent series of forty standard thin sections features important and interesting rock-forming mineral types most often found in field samples.
Each thin section is made from pure mineral samples or from rocks containing a good amount of the mineral. This set is housed in standard slide boxes and an identification list is included.
Specifications
- Description:Mineral Thin Sections Set
- No. of Slides:40