Orders LinkContactUsLinkComponent
Polymethylpentene Cylinders
 :   DYNALON
Polymethylpentene Cylinders
 :  DYNALON
Polymethylpentene Cylinders
Polymethylpentene Cylinders
These polymethylpentene cylinders provide glass-like clarity and can withstand temperatures up to 350°F.
  • Heat-resistant
  • Shatterproof
The cylinders may be autoclaved, and they are chemically resistant to most acids, bases, alcohols, and aldehydes. All cylinders are calibrated to contain.
  • Catalog No:
  • 470174-192
  • 470174-186
  • Form:
  • Tall
Product Family Options
  • Capacity
    Graduation Interval
  • 250 ml
    2.0 ml
    Each (1UN)
    Each (1UN) - $307.40
    $307.40
    Case de 8 (1UN)
    Case de 8 (1UN) - $335.07
    $335.07
    Each (1UN)
       $307.40
    Case de 8 (1UN)
       $335.07
     : 237775
    Specifications
    Cat. No.
    470174-192
    Form
    Tall
  • 1000 ml
    10.0 ml
    Each (1UN)
    Each (1UN) - $759.95
    $759.95
    Case de 4 (1UN)
    Case de 4 (1UN) - $828.34
    $828.34
    Each (1UN)
       $759.95
    Case de 4 (1UN)
       $828.34
     : 237895
    Specifications
    Cat. No.
    470174-186
    Form