Orders LinkContactUsLinkComponent
Polypropylene Cylinders
Polypropylene Cylinders
Polypropylene Cylinders
Polypropylene Cylinders
Unbreakable Polypropylene.
  • Permanent, molded, single scale metric markings
  • Flared top rims and large spouts facilitate pouring
  • Hexagonal shaped bases prevent tipping and rolling
  • Autoclavable at 121°C
Chemically resistant to most acids, bases, alcohols, esters, and ketones.
Product Family Options
  • Capacity
    Ext.Ø
    Graduation Interval
  • 10 ml
    12.50 mm
    0.2 ml
    Each (1UN)
    Each (1UN) - $9.68
    $9.68
    Case de 24 (1UN)
    Case de 24 (1UN) - $10.55
    $10.55
    Each (1UN)
       $9.68
    Case de 24 (1UN)
       $10.55
  • 25 ml
    17.20 mm
    0.5 ml
    Each (1UN)
    Each (1UN) - $12.36
    $12.36
    Case de 24 (1UN)
    Case de 24 (1UN) - $13.47
    $13.47
    Each (1UN)
       $12.36
    Case de 24 (1UN)
       $13.47
  • 50 ml
    24.25 mm
    1.0 ml
    Each (1UN)
    Each (1UN) - $16.11
    $16.11
    Case de 24 (1UN)
    Case de 24 (1UN) - $17.56
    $17.56
    Each (1UN)
       $16.11
    Case de 24 (1UN)
       $17.56
  • 100 ml
    27.00 mm
    1.0 ml
    Each (1UN)
    Each (1UN) - $20.97
    $20.97
    Case de 12 (1UN)
    Case de 12 (1UN) - $22.86
    $22.86
    Each (1UN)
       $20.97
    Case de 12 (1UN)
       $22.86
  • 250 ml
    40.60 mm
    2.0 ml
    Each (1UN)
    Each (1UN) - $38.70
    $38.70
    Case de 6 (1UN)
    Case de 6 (1UN) - $42.18
    $42.18
    Each (1UN)
       $38.70
    Case de 6 (1UN)
       $42.18
  • 500 ml
    52.50 mm
    5.0 ml
    Each (1UN)
    Each (1UN) - $59.12
    $59.12
    Case de 6 (1UN)
    Case de 6 (1UN) - $64.44
    $64.44
    Each (1UN)
       $59.12
    Case de 6 (1UN)
       $64.44
  • 1000 ml
    _
    10 ml
    Each (1UN)
    Each (1UN) - $261.00
    $261.00
    Case de 6 (1UN)
    Case de 6 (1UN) - $309.88
    $309.88
    Each (1UN)
       $261.00
    Case de 6 (1UN)
       $309.88