Urine Test Strips
Supplier: LW SCIENTIFIC SE
Jump to:
About this item
Easy-to-use test strips check for up to 10 parameters.
- Easy to Use
5 Parameter Urine Reagent Strips: Glucose, Protein, pH, Blood, Ketone
10 Parameter Urine Reagent Strips: Glucose, Protein, pH, Blood, Ketone, bilirubin, nitrite, urobilinogen, specific gravity, and leukocytes.
2 Options Available Below
Product Details & Documents
Easy-to-use test strips check for up to 10 parameters.
- Easy to Use
5 Parameter Urine Reagent Strips: Glucose, Protein, pH, Blood, Ketone
10 Parameter Urine Reagent Strips: Glucose, Protein, pH, Blood, Ketone, bilirubin, nitrite, urobilinogen, specific gravity, and leukocytes.
Product Family Options
Product Information
- AvailabilityPrice
- 470101-700Urine Test Strips, 5 Parameters Pk/100Supplier Number: URS-05PR-7777
- 470210-690Urine Test Strips, 10 ParametersSupplier Number: URS-10PR-7777