Flexible Feel Inoculation Loops in Tamper-Evident Resealable Bags, Globe Scientific
Catalog # 10799-759
Supplier: GLOBE SCIENTIFIC INC
Specifications
- Esterilidad:Sterile
- Capacidad:10 µl
- Referencia:10799-759
- Empaquetado:10/Zip Lock Bag, 100 Bags/Case
- Nº de proveedor:2875-10
Specifications
About this item
These disposable inoculation loops and needles offer the most convenient and safest way of streaking culture media.
Gamma sterilized, sterility assurance level 10-3
- Certificate of calibration
- Tamper-evident resealable bags
- Integrated loop/needle design
The loops are packaged in tamper-evident resealable bags. All loops are sterilized by gamma irradiation. The 1µl and 10µl inoculation loops are available with the loop on one end and an integrated needle on the other end, eliminating the need for a separate needle.
The polystyrene (PS) inoculation loops are smooth and flexible to facilitate uniform streaking of culture media.
Certifications: Manufactured under ISO 9001:2008 and ISO 14001:2004 standards