Orders LinkContactUsLinkComponent
Graduated Measure, Sklar
Graduated Measure, Sklar
32 oz., 133 mm (5¼"), 121 mm (4¾")
Catalog # 10163-848
Supplier:   SKLAR CORPORATION
Supplier #:  10-1510
Graduated Measure, Sklar
Catalog # 10163-848
Supplier:  SKLAR CORPORATION
Graduated Measure, Sklar
Graduated Measure, Sklar
32 oz., 133 mm (5¼"), 121 mm (4¾")
Price
$5,570.87
Log in Para disponibilidad
Each (1UN)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Sklar's® Graduated Measure is a container used to accurately measure fluids or measure and dilute chemicals for use
It resembles a measuring cup and may be referred to as simply a "measure" or a "graduate." Calibrations for cubic centimeters and ounces are indicated on the inside of the product. Stainless steel.
Especificaciones
  • Ext.Ø:
    121 mm (4¾")
  • Capacity:
    32 oz.
  • Height:
    133 mm (5¼")
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 