PYREX® Pennyhead Stopper, Corning
13
Catalog # 89134-418
Product available on GSA Advantage®
Supplier: Corning
Número de proveedor #: 7661-13
PYREX® Pennyhead Stopper, Corning
Catalog # 89134-418
Supplier: Corning
Product available on GSA Advantage®
Jump to:
Product Details & Documents
PYREX® glass pennyhead stoppers are generally used in flasks and separatory funnels.
Especificaciones
- ST Stopper No.:13
- Cat. No.:89134-418
- Environmental certification:I-RECs, ISO 14001 - Monterrey
- Environmentally Preferable:Low Manufacturing Impact