ContactUsLinkComponent
PYREX® 75° Angled Tube Adapter with Two-Way 24/40 Standard Taper Joints, Corning
Catalog # 89091-524
Supplier:  Corning
Product available on GSA Advantage®
   Low Manufacturing Impact
undefined
PYREX® 75° Angled Tube Adapter with Two-Way 24/40 Standard Taper Joints, Corning
Catalog # 89091-524
Supplier:  Corning
Supplier Number:  8920-24
Product available on GSA Advantage®
   Low Manufacturing Impact
Specifications
  • Descripción:
    PYREX® 75° Angled Tube Adapter with Two-Way [ST] Joints
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Referencia:
    89091-524
  • ST Joint Size:
    24/40
  • Nº de proveedor:
    8920-24
  • Certificación ambiental:
    I-RECs, ISO 14001 - Monterrey
Specifications
About this item
These angled two-way adapters are made of PYREX® borosilicate glass.
  • Arms are approximately 75° apart
  • Both ends are equipped with an inner 24/40 Standard Taper joint
It is often used as a stillhead adapter.