PYREX® Blunt Tip Imhoff Sediment Cone, Corning
Imhoff Sediment Cone, Blunt Tip, PYREX®, 1000 ml, 435 mm, 110 mm
Supplier: Corning
Número de proveedor #: 2180-439
PYREX® Blunt Tip Imhoff Sediment Cone, Corning
Catalog # 89091-192
Supplier: Corning
Product available on GSA Advantage®
Jump to:
Product Details & Documents
Made of PYREX® borosilicate glass.
- Also graduated and marked at 1000 ml
- Blunt tip increases ruggedness and cleaning ease
- Very good chemical and temperature resistance
These blunt tip cones are used with heavy sediments, where ability to read small volumes is unimportant. Graduated from 0 ml to 1 ml in 0.1 ml increments, 1 to 10 ml in 0.5 ml increments and 10 ml to 40 ml in 1 ml increments.
Documents
Certificates
Buscar por número de loteEspecificaciones
- Capacity:1000 ml
- Description:Imhoff Sediment Cone, Blunt Tip, PYREX®
- Height:435 mm
- Top I.D.:110 mm
- Cat. No.:89091-192
- Environmental certification:I-RECs, ISO 14001 - Monterrey
- Environmentally Preferable:Low Manufacturing Impact