Corning® Disposable Phenolic Threaded Screw Cap with Rubber Liners, Corning
Supplier: Corning
Certificates
About this item
These disposable Corning® caps are made from phenolic resin and are autoclavable at 121°C
- Available in different sizes
They are provided with a white rubber liner.
Specifications
- Catalog No:
- 60826-483
- 60826-494
- 60827-110
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Certificación ambiental:
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey