Corning® Polypropylene Funnel, Corning
Short Stem Funnel, 150 mm, PP, 220 mm
: Corning
: 6120P-150
Funnels, made of durable PP, translucent.
- Very good chemical resistance
- Autoclavable
- Suitable for use with standard filter paper
Allow safe and easy transfers of solids or liquids from one vessel to another.
- Ø:150 mm
- Description:Short Stem Funnel
- Length:220 mm
- Material:PP