Orders LinkContactUsLinkComponent
PYREX® Reaction Vial, Corning
PYREX® Reaction Vial, Corning
Supplier:   Corning
PYREX® Reaction Vial, Corning
Supplier:  Corning
Certificates
PYREX® Reaction Vial, Corning
PYREX® Reaction Vial, Corning
3 Options Available Below
Product Details & Documents
Conical Reaction Vials serve as a replacement part for the 6949M series of PYREX® Microchemistry Kits.
Documents
Product Family Options
Product Information
  • Capacity
    Bottom Style
    Material
    Cap included
    Availability
    Price
  • 89091-556
    Reaction vial
    Reaction vial
    0.10 ml
    Conical bottom
    PYREX® glass
    Yes
    Log in for availability
    Log in for availability
    Case (12UN)
    Case (12UN) - $14,650.00
    $14,650.00
    Case (12UN)
       $14,650.00
    Supplier Number: 9900-1X
  • 89091-562
    Reaction vial
    Reaction vial
    3.00 ml
    Conical bottom
    PYREX® glass
    Yes
    Log in for availability
    Log in for availability
    Case (12UN)
    Case (12UN) - $13,200.00
    $13,200.00
    Case (12UN)
       $13,200.00
    Supplier Number: 9900-3
  • Reaction vial
    5.00 ml
    Conical bottom
    PYREX® glass
    Yes
    Log in for availability
    Log in for availability
    Case (12UN)
    Case (12UN) - $13,200.00
    $13,200.00
    Case (12UN)
       $13,200.00
    Supplier Number: 9900-5