PYREX® Liebig Condensers, with Drip Tip and [ST] Outer and Inner Joints, Corning
Proveedor: Corning
Certificados
Acerca de este artículo
Made of PYREX® borosilicate glass.
- With outer joint at the top and inner standard taper joint with drip tip
- Very good temperature resistance
- Very good chemical resistance
This general purpose condenser can be used for distillation, refluxing and extraction operations. The condenser tubulations are approximately 10 mm Ext.Ø. Can be used for vacuum distillation.
Especificaciones
- Catalog No:
- 89090-726
- 89091-374
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Certificación ambiental:
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey