PYREX® Chromatography Column with Coarse Fritted Disc, Corning
89091-236
: Corning
Product available on GSA Advantage®
- Ø int.:22.00 mm
- Descripción:PYREX® Chromatography Column with Coarse Fritted Disc
- Longitud:400 mm
- Environmentally Preferable:Low Manufacturing Impact
- Certificación ambiental:I-RECs, ISO 14001 - Monterrey
- Referencia:89091-236
- Stopper No.:4
- Nº de proveedor:38450-20
These columns are designed for general chromatography analysis.
The tops are tooled for rubber stopper fit and the tubes are fabricated with a sealed-in coarse porosity fritted disc.